Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201743.3 | NP_001188672.1 | 4 - 5 | 1592 | 104 | |
NM_001258827.1 | NP_001245756.1 | 4 - 5 | 1532 | 104 | ||
NM_001272779.1 | NP_001259708.1 | 4 - 5 | 1787 | 104 | ||
NM_001272780.2 | NP_001259709.1 | 4 - 5 | 1592 | 104 | ||
NM_167665.2 | NP_728267.1 | 4 - 5 | 1242 | 104 | ||
NM_167666.3 | NP_728268.1 | 4 - 5 | 1592 | 104 | ||
GenBank mRNA | AB277547.1 | - | 4 - 5 | 1571 | 104 | |
BT099719.1 | - | 4 - 5 | 1531 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |