Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mthl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_167821.3 | NP_728509.1 | 7 - 8 | 1498 | 99 | |
GenBank mRNA | AY071287.1 | - | 7 - 8 | 1494 | 99 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Drosophila melanogaster |