Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144390.2 | NP_001137862.1 | 4 - 5 | 1734 | 118 | |
NM_001144391.1 | NP_001137863.1 | 2 - 3 | 1647 | 118 | ||
NM_001169836.1 | NP_001163307.1 | 4 - 5 | 1471 | 118 | ||
NM_001169837.1 | NP_001163308.1 | 4 - 5 | 1546 | 118 | ||
NM_001274273.1 | NP_001261202.1 | 13 - 14 | 9500 | 118 | ||
NM_001382119.1 | NP_001368935.1 | 3 - 4 | 1598 | 118 | ||
NM_138180.4 | NP_612024.4 | 13 - 14 | 9503 | 118 | ||
NM_138182.2 | NP_612026.2 | 4 - 5 | 1890 | 118 | ||
NM_138183.3 | NP_612027.2 | 4 - 5 | 1734 | 118 | ||
NM_167823.2 | NP_728515.1 | 4 - 5 | 1563 | 118 | ||
NM_167824.2 | NP_728516.1 | 4 - 5 | 1557 | 118 | ||
NM_167825.2 | NP_728517.1 | 3 - 4 | 1542 | 118 | ||
GenBank mRNA | BT044233.1 | - | 4 - 5 | 1714 | 118 | |
BT099707.1 | - | 4 - 5 | 1890 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |