Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_167824.2 | NP_728516.1 | 1 - 2 | 280 | 73 | |
GenBank mRNA | BT016166.1 | - | 1 - 2 | 239 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |