Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272173.1 | NP_001259102.1 | 9 - 10 | 2263 | 73 | |
NM_130502.3 | NP_569858.1 | 9 - 10 | 2263 | 73 | ||
GenBank mRNA | AB176551.1 | - | 9 - 10 | 2104 | 73 | |
AY095525.1 | - | 9 - 10 | 2218 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |