Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201609.1 | NP_001188538.1 | 2 - 3 | 285 | 99 | |
NM_001201610.2 | NP_001188539.1 | 2 - 3 | 592 | 99 | ||
NM_001272256.1 | NP_001259185.1 | 2 - 3 | 353 | 99 | ||
NM_130644.4 | NP_570000.1 | 2 - 3 | 1559 | 99 | ||
GenBank mRNA | BT088778.1 | - | 2 - 3 | 405 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |