Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trol | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031864.4 | NP_001027035.3 | 5 - 6 | 1202 | 96 | |
NM_001031866.4 | NP_001027037.3 | 5 - 6 | 1202 | 96 | ||
NM_001169177.4 | NP_001162648.3 | 5 - 6 | 1202 | 96 | ||
NM_001258563.3 | NP_001245492.2 | 5 - 6 | 1202 | 96 | ||
NM_001258564.3 | NP_001245493.2 | 5 - 6 | 1202 | 96 | ||
NM_001258567.3 | NP_001245496.2 | 5 - 6 | 1202 | 96 | ||
NM_001258568.3 | NP_001245497.2 | 5 - 6 | 1202 | 96 | ||
NM_001258569.3 | NP_001245498.2 | 5 - 6 | 1202 | 96 | ||
NM_001258570.3 | NP_001245499.2 | 5 - 6 | 1202 | 96 | ||
NM_001258571.3 | NP_001245500.2 | 5 - 6 | 1202 | 96 | ||
NM_001297893.2 | NP_001284822.1 | 5 - 6 | 1202 | 96 | ||
NM_001297896.2 | NP_001284825.1 | 5 - 6 | 1202 | 96 | ||
NM_001382068.1 | NP_001368970.1 | 5 - 6 | 1202 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |