Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pirk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299757.1 | NP_001286686.1 | 2 | 1772 | 100 | |
NM_137754.2 | NP_611598.1 | 1 | 1227 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |