Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299771.1 | NP_001286700.1 | 5 - 6 | 1402 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |