Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GlcT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137792.3 | NP_611636.1 | 1 | 1646 | 65 | |
GenBank mRNA | AY118335.1 | - | 1 | 1592 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |