Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oatp58Dc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103939.2 | NP_001097409.1 | 3 - 4 | 1061 | 87 | |
NM_001169780.2 | NP_001163251.1 | 2 - 3 | 841 | 87 | ||
NM_137815.3 | NP_611659.2 | 3 - 4 | 1103 | 87 | ||
GenBank mRNA | BT023942.1 | - | 2 - 3 | 1820 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |