Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obp58b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137865.3 | NP_611709.1 | 1 - 2 | 55 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |