Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obp58d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137867.2 | NP_611711.1 | 2 - 3 | 272 | 85 | |
GenBank mRNA | BT128702.1 | - | 2 - 3 | 258 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |