Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138015.4 | NP_611859.1 | 2 - 3 | 476 | 93 | |
GenBank mRNA | AY051603.1 | - | 2 - 3 | 457 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |