Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snama | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259558.2 | NP_001246487.1 | 4 - 5 | 1125 | 126 | |
NM_138040.4 | NP_611884.1 | 4 - 5 | 997 | 126 | ||
GenBank mRNA | AF132177.1 | - | 4 - 5 | 969 | 126 | |
BT124892.1 | - | 4 - 5 | 1335 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |