Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nord | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138056.3 | NP_611900.1 | 2 - 3 | 691 | 70 | |
GenBank mRNA | BT133485.1 | - | 2 - 3 | 683 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |