Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299917.1 | NP_001286846.1 | 1 - 2 | 706 | 99 | |
NM_138090.2 | NP_611934.1 | 1 - 2 | 545 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |