Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143665.4 | NP_651922.1 | 2 - 3 | 989 | 100 | |
GenBank mRNA | AY051651.1 | - | 2 - 3 | 918 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |