Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143665.4 | NP_651922.1 | 3 - 4 | 1200 | 71 | |
GenBank mRNA | AY051651.1 | - | 3 - 4 | 1129 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |