Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NtR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259528.2 | NP_001246457.1 | 2 - 3 | 504 | 69 | |
NM_143701.4 | NP_651958.2 | 2 - 3 | 504 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |