Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274330.1 | NP_001261259.1 | 18 - 19 | 4153 | 103 | |
NM_001274331.1 | NP_001261260.1 | 18 - 19 | 4129 | 103 | ||
NM_139361.2 | NP_647618.1 | 17 - 18 | 4033 | 103 | ||
NM_167888.2 | NP_728620.1 | 17 - 18 | 4036 | 103 | ||
NM_167889.2 | NP_728621.1 | 19 - 20 | 4252 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |