Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dre4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057262.4 | NP_476610.2 | 3 - 4 | 766 | 109 | |
NM_167922.3 | NP_728686.2 | 3 - 4 | 766 | 109 | ||
GenBank mRNA | BT010116.1 | - | 3 - 4 | 790 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |