Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: promL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259658.3 | NP_001246587.1 | 9 - 10 | 1855 | 68 | |
NM_001274422.1 | NP_001261351.1 | 8 - 9 | 1772 | 68 | ||
NM_001274424.1 | NP_001261353.1 | 8 - 9 | 1974 | 68 | ||
NM_001382193.1 | NP_001368963.1 | 9 - 10 | 1855 | 68 | ||
NM_139513.3 | NP_647770.1 | 8 - 9 | 1696 | 68 | ||
NM_167986.1 | NP_728809.1 | 8 - 9 | 1974 | 68 | ||
NM_167987.4 | NP_728810.1 | 9 - 10 | 1855 | 68 | ||
GenBank mRNA | AF127935.2 | - | 9 - 10 | 1830 | 68 | |
AF197345.1 | - | 8 - 9 | 1652 | 68 | ||
BT031128.1 | - | 8 - 9 | 1654 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |