Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ens | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104028.2 | NP_001097498.1 | 5 - 6 | 655 | 57 | |
NM_001274470.1 | NP_001261399.1 | 4 - 5 | 538 | 57 | ||
NM_001274471.1 | NP_001261400.1 | 4 - 5 | 538 | 57 | ||
NM_001274472.1 | NP_001261401.1 | 4 - 5 | 538 | 57 | ||
NM_001274473.1 | NP_001261402.1 | 4 - 5 | 538 | 57 | ||
NM_001274474.1 | NP_001261403.1 | 4 - 5 | 538 | 57 | ||
NM_001300004.1 | NP_001286933.1 | 4 - 5 | 547 | 57 | ||
NM_139604.2 | NP_647861.2 | 3 - 4 | 493 | 57 | ||
NM_168061.2 | NP_728938.2 | 4 - 5 | 547 | 57 | ||
NM_168062.3 | NP_728939.2 | 4 - 5 | 547 | 57 | ||
NM_168063.2 | NP_728940.2 | 4 - 5 | 547 | 57 | ||
NM_168064.2 | NP_728941.2 | 5 - 6 | 655 | 57 | ||
GenBank mRNA | BT010301.1 | - | 5 - 6 | 613 | 57 | |
BT125906.1 | - | 4 - 5 | 533 | 57 | ||
BT126155.1 | - | 4 - 5 | 528 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |