Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: loj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139770.4 | NP_648027.3 | 4 - 5 | 601 | 83 | |
NM_168154.3 | NP_729140.2 | 3 - 4 | 696 | 83 | ||
NM_168155.3 | NP_729141.2 | 4 - 5 | 621 | 83 | ||
GenBank mRNA | AY061384.1 | - | 3 - 4 | 576 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |