Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dikar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274552.1 | NP_001261481.1 | 6 - 7 | 2469 | 62 | |
NM_168179.3 | NP_729188.2 | 6 - 7 | 2469 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |