Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dikar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202125.2 | NP_001189054.1 | 8 - 9 | 3167 | 98 | |
NM_001274551.1 | NP_001261480.1 | 8 - 9 | 3167 | 98 | ||
NM_001274552.1 | NP_001261481.1 | 8 - 9 | 2741 | 98 | ||
NM_168179.3 | NP_729188.2 | 8 - 9 | 2741 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |