Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dikar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202125.2 | NP_001189054.1 | 9 - 10 | 7316 | 72 | |
NM_001274551.1 | NP_001261480.1 | 9 - 10 | 7316 | 72 | ||
NM_001274552.1 | NP_001261481.1 | 9 - 10 | 6890 | 72 | ||
NM_168179.3 | NP_729188.2 | 9 - 10 | 6890 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |