Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGEF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139844.3 | NP_648101.1 | 3 - 4 | 1343 | 75 | |
NM_168199.2 | NP_729232.1 | 2 - 3 | 1144 | 75 | ||
GenBank mRNA | AF255674.1 | - | 3 - 4 | 1341 | 75 | |
AY058496.1 | - | 3 - 4 | 1312 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |