Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CheA7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132167.3 | NP_572395.2 | 1 - 2 | 251 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |