Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir7e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144704.1 | NP_001138176.1 | 1 - 2 | 815 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |