Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298064.1 | NP_001284993.1 | 7 - 8 | 2759 | 87 | |
NM_132193.2 | NP_572421.1 | 7 - 8 | 2759 | 87 | ||
GenBank mRNA | BT150246.1 | - | 7 - 8 | 2526 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |