Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201630.2 | NP_001188559.1 | 5 - 6 | 2071 | 79 | |
NM_001201631.2 | NP_001188560.1 | 6 - 7 | 1872 | 79 | ||
NM_132230.4 | NP_572458.3 | 5 - 6 | 2071 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |