Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cp7Fc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132257.2 | NP_572485.1 | 1 - 2 | 82 | 56 | |
GenBank mRNA | BT031227.1 | - | 1 - 2 | 82 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |