Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mthl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274253.1 | NP_001261182.1 | 2 - 3 | 2213 | 87 | |
NM_138154.3 | NP_611998.1 | 1 - 2 | 529 | 87 | ||
GenBank mRNA | AY069723.1 | - | 1 - 2 | 1755 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |