Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mwh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274314.1 | NP_001261243.1 | 8 - 9 | 2267 | 77 | |
NM_001274315.1 | NP_001261244.1 | 8 - 9 | 2270 | 77 | ||
NM_138238.3 | NP_612082.1 | 6 - 7 | 1758 | 77 | ||
GenBank mRNA | AM231270.1 | - | 6 - 7 | 1743 | 77 | |
BT023932.1 | - | 6 - 7 | 1755 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |