Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SA-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138265.3 | NP_612109.3 | 2 - 3 | 621 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |