Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169916.2 | NP_001163387.2 | 8 - 9 | 4511 | 65 | |
NM_001202142.2 | NP_001189071.1 | 6 - 7 | 1094 | 65 | ||
NM_001202143.2 | NP_001189072.1 | 6 - 7 | 1094 | 65 | ||
NM_001259753.2 | NP_001246682.1 | 5 - 6 | 773 | 65 | ||
NM_001259754.2 | NP_001246683.1 | 6 - 7 | 1094 | 65 | ||
NM_001259755.1 | NP_001246684.1 | 6 - 7 | 2096 | 65 | ||
NM_001274670.1 | NP_001261599.1 | 8 - 9 | 5035 | 65 | ||
NM_168293.3 | NP_729410.1 | 6 - 7 | 1094 | 65 | ||
NM_168294.3 | NP_729411.1 | 6 - 7 | 1094 | 65 | ||
GenBank mRNA | BT050456.1 | - | 6 - 7 | 1060 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |