Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbfox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104102.4 | NP_001097572.1 | 7 - 8 | 1898 | 69 | |
NM_001104103.3 | NP_001097573.1 | 7 - 8 | 1883 | 69 | ||
NM_001259779.1 | NP_001246708.1 | 7 - 8 | 2406 | 69 | ||
NM_168412.4 | NP_729614.2 | 8 - 9 | 2081 | 69 | ||
GenBank mRNA | BT010058.1 | - | 7 - 8 | 1870 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |