Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rols | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274807.1 | NP_001261736.1 | 5 - 6 | 1598 | 58 | |
NM_168487.2 | NP_729778.1 | 5 - 6 | 2027 | 58 | ||
NM_168489.2 | NP_729780.1 | 5 - 6 | 1584 | 58 | ||
NM_168490.2 | NP_729781.1 | 5 - 6 | 1601 | 58 | ||
GenBank mRNA | AF440568.1 | - | 4 - 5 | 1157 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |