Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp68C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168558.2 | NP_729926.1 | 2 | 1122 | 88 | |
NM_168559.2 | NP_729927.2 | 2 | 1122 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |