Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104144.3 | NP_001097614.1 | 3 - 4 | 860 | 88 | |
NM_001104145.3 | NP_001097615.1 | 3 - 4 | 853 | 88 | ||
NM_001259854.2 | NP_001246783.1 | 3 - 4 | 1110 | 88 | ||
NM_001259855.2 | NP_001246784.1 | 3 - 4 | 884 | 88 | ||
NM_001259856.2 | NP_001246785.1 | 3 - 4 | 1134 | 88 | ||
NM_001259857.2 | NP_001246786.1 | 3 - 4 | 884 | 88 | ||
NM_001259858.2 | NP_001246787.1 | 3 - 4 | 860 | 88 | ||
NM_001274990.1 | NP_001261919.1 | 3 - 4 | 860 | 88 | ||
NM_001382059.1 | NP_001369057.1 | 3 - 4 | 860 | 88 | ||
NM_168646.3 | NP_730099.2 | 3 - 4 | 853 | 88 | ||
NM_168647.3 | NP_730100.2 | 3 - 4 | 860 | 88 | ||
NM_168648.3 | NP_730101.2 | 3 - 4 | 835 | 88 | ||
GenBank mRNA | AF500094.1 | - | 2 - 3 | 367 | 88 | |
BT133053.1 | - | 3 - 4 | 841 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |