Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104144.3 | NP_001097614.1 | 6 - 7 | 1613 | 59 | |
NM_001104145.3 | NP_001097615.1 | 6 - 7 | 1609 | 59 | ||
NM_001259854.2 | NP_001246783.1 | 6 - 7 | 1863 | 59 | ||
NM_001259856.2 | NP_001246785.1 | 6 - 7 | 1890 | 59 | ||
NM_001259857.2 | NP_001246786.1 | 6 - 7 | 1637 | 59 | ||
NM_001259858.2 | NP_001246787.1 | 7 - 8 | 2052 | 59 | ||
NM_001274990.1 | NP_001261919.1 | 6 - 7 | 1613 | 59 | ||
NM_001382059.1 | NP_001369057.1 | 6 - 7 | 1613 | 59 | ||
NM_168646.3 | NP_730099.2 | 6 - 7 | 1606 | 59 | ||
NM_168647.3 | NP_730100.2 | 7 - 8 | 1808 | 59 | ||
NM_168648.3 | NP_730101.2 | 6 - 7 | 1588 | 59 | ||
GenBank mRNA | AF500094.1 | - | 5 - 6 | 1120 | 59 | |
BT133053.1 | - | 6 - 7 | 1594 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |