Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ome | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104136.2 | NP_001097606.1 | 10 - 11 | 3182 | 83 | |
NM_001144483.2 | NP_001137955.1 | 9 - 10 | 3221 | 83 | ||
NM_001274915.1 | NP_001261844.1 | 8 - 9 | 2375 | 83 | ||
NM_080000.4 | NP_524739.1 | 9 - 10 | 2468 | 83 | ||
GenBank mRNA | AY058514.1 | - | 9 - 10 | 2441 | 83 | |
BT024224.1 | - | 9 - 10 | 3192 | 83 | ||
BT029957.1 | - | 10 - 11 | 3166 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |