Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272879.1 | NP_001259808.1 | 3 - 4 | 403 | 85 | |
NM_080010.4 | NP_524749.1 | 3 - 4 | 403 | 85 | ||
GenBank mRNA | AY075315.1 | - | 3 - 4 | 349 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |