Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: futsch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103394.3 | NP_001096864.1 | 4 - 5 | 1373 | 67 | |
NM_001169161.3 | NP_001162632.2 | 5 - 6 | 2009 | 67 | ||
NM_001258532.1 | NP_001245461.1 | 5 - 6 | 2809 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |