Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GluRIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274692.1 | NP_001261621.2 | 9 - 10 | 2156 | 67 | |
NM_001274693.1 | NP_001261622.1 | 9 - 10 | 2156 | 67 | ||
NM_001316480.1 | NP_001303409.1 | 9 - 10 | 2156 | 67 | ||
NM_080016.4 | NP_524755.2 | 9 - 10 | 2156 | 67 | ||
GenBank mRNA | BT150463.1 | - | 9 - 10 | 2155 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |