Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258636.2 | NP_001245565.1 | 4 - 5 | 1835 | 148 | |
NM_080025.2 | NP_524764.1 | 3 - 4 | 2075 | 148 | ||
GenBank mRNA | X07985.1 | - | 3 - 4 | 2075 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |