Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nkd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080049.3 | NP_524788.2 | 3 - 4 | 2430 | 54 | |
GenBank mRNA | BT015258.1 | - | 3 - 4 | 2395 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |