Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ric8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298119.1 | NP_001285048.1 | 1 - 2 | 215 | 62 | |
NM_132322.4 | NP_572550.2 | 1 - 2 | 215 | 62 | ||
GenBank mRNA | BT004873.1 | - | 1 - 2 | 192 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |